ENA432538250 Enamine

(2S)-2-[(4-chlorophenyl)amino]propanoic acid

  • MDL number MFCD20641844
  • Cas Number : 103678-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192033
Availability details for
Quantity
  • Availability
  • Add to Cart