ENA432538251 Enamine

methyl 2-(benzyloxy)propanoate

  • MDL number MFCD00792541
  • Cas Number : 53346-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134465
Availability details for
Quantity
  • Availability
  • Add to Cart