ENA432538254 Enamine

N-(2-hydroxyethyl)-4-methoxybenzamide

  • MDL number MFCD00466253
  • Cas Number : 57728-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192148
Availability details for
Quantity
  • Availability
  • Add to Cart