ENA432538255 Enamine

4-(2-fluorophenyl)-1,2-oxazol-3-amine

  • MDL number MFCD07382300
  • Cas Number : 1267666-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192160
Availability details for
Quantity
  • Availability
  • Add to Cart