ENA432538256 Enamine

5-methyl-2,3-dihydro-1H-isoindole

  • MDL number MFCD09056876
  • Cas Number : 93282-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-44
Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192161
Availability details for
Quantity
  • Availability
  • Add to Cart