ENA432538257 Enamine

5-chloro-2-methyl-1,3-benzothiazol-4-amine

  • MDL number MFCD20703588
  • Cas Number : 855463-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-109
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192228
Availability details for
Quantity
  • Availability
  • Add to Cart