ENA432538258 Enamine

N,N-dimethyl-4-(pyridin-4-yl)aniline

  • MDL number MFCD00186957
  • Cas Number : 1137-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231-236
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134516
Availability details for
Quantity
  • Availability
  • Add to Cart