ENA432538263 Enamine

3-(ethylsulfanyl)pyrazine-2-carbonitrile

  • MDL number MFCD16843442
  • Cas Number : 74002-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192396
Availability details for
Quantity
  • Availability
  • Add to Cart