ENA432538266 Enamine

1,3-dibromo-2-(2-bromoethoxy)benzene

  • MDL number MFCD11119884
  • Cas Number : 206347-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192444
Availability details for
Quantity
  • Availability
  • Add to Cart