ENA432538267 Enamine

4-bromo-6-[(4-methoxyphenyl)methoxy]-2-methylpyrimidine

  • MDL number MFCD27814040
  • Cas Number : 1715298-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192455
Availability details for
Quantity
  • Availability
  • Add to Cart