ENA432538268 Enamine

2-methanesulfonylpropan-1-ol

  • MDL number MFCD26797775
  • Cas Number : 21975-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192467
Availability details for
Quantity
  • Availability
  • Add to Cart