ENA432538270 Enamine

7-bromo-2-methylimidazo[1,2-a]pyridine

  • MDL number MFCD11226901
  • Cas Number : 1194375-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192563
Availability details for
Quantity
  • Availability
  • Add to Cart