ENA432538278 Enamine

1,3-diethyl 2-(3-chlorophenyl)-4-hydroxy-4-methyl-6-oxocyclohexane-1,3-dicarboxylate

  • MDL number MFCD00785410
  • Cas Number : 4759-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192630
Availability details for
Quantity
  • Availability
  • Add to Cart