ENA432538283 Enamine

butyl 2-fluoro-6-hydroxybenzoate

  • MDL number MFCD28139412
  • Cas Number : 1056955-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192644
Availability details for
Quantity
  • Availability
  • Add to Cart