ENA432538284 Enamine

3-(4-bromophenyl)-5-ethyl-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD19092951
  • Cas Number : 1463808-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192659
Availability details for
Quantity
  • Availability
  • Add to Cart