ENA432538288 Enamine

ethyl 2-[(4-methylphenyl)amino]acetate

  • MDL number MFCD11154309
  • Cas Number : 21911-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192680
Availability details for
Quantity
  • Availability
  • Add to Cart