ENA432538292 Enamine

(2R)-2-amino-1-(pyrrolidin-1-yl)propan-1-one

  • MDL number MFCD16051072
  • Cas Number : 1152110-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192699
Availability details for
Quantity
  • Availability
  • Add to Cart