ENA432538300 Enamine

2-(chloromethyl)benzene-1-sulfonamide

  • MDL number MFCD24459680
  • Cas Number : 81629-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192785
Availability details for
Quantity
  • Availability
  • Add to Cart