ENA432538302 Enamine

5-nitropyridine-2-carbaldehyde

  • MDL number MFCD07368185
  • Cas Number : 35969-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-68
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-137819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137819
Availability details for
Quantity
  • Availability
  • Add to Cart