ENA432538332 Enamine

2-benzyl-5-bromo-1,3,4-oxadiazole

  • MDL number MFCD21892330
  • Cas Number : 1369032-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-192957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192957
Availability details for
Quantity
  • Availability
  • Add to Cart