ENA432538334 Enamine

2-methyl-5-(pyridin-3-yl)aniline

  • MDL number MFCD12779835
  • Cas Number : 1007213-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192962
Availability details for
Quantity
  • Availability
  • Add to Cart