ENA432538336 Enamine

4-methoxy-4-methylazepane hydrochloride

  • MDL number MFCD28139429
  • Cas Number : 1795535-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192965
Availability details for
Quantity
  • Availability
  • Add to Cart