ENA432538339 Enamine

2-phenylpyrimidine-4,6-diamine

  • MDL number MFCD18819665
  • Cas Number : 52644-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192970
Availability details for
Quantity
  • Availability
  • Add to Cart