ENA432538340 Enamine

2-chloro-6-methylpyrimidine-4-carbonitrile

  • MDL number MFCD09878518
  • Cas Number : 16858-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192972
Availability details for
Quantity
  • Availability
  • Add to Cart