ENA432538341 Enamine

2-(prop-2-yn-1-yloxy)-6-(trifluoromethyl)pyridine

  • MDL number MFCD28139432
  • Cas Number : 1787995-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192983
Availability details for
Quantity
  • Availability
  • Add to Cart