ENA432538343 Enamine

{5-[(dimethylamino)methyl]thiophen-2-yl}methanamine

  • MDL number MFCD06803279
  • Cas Number : 893742-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192985
Availability details for
Quantity
  • Availability
  • Add to Cart