ENA432538348 Enamine

2-methoxy-3-methylpentanoic acid

  • MDL number MFCD24499319
  • Cas Number : 1546711-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193000
Availability details for
Quantity
  • Availability
  • Add to Cart