ENA432538351 Enamine

2-isocyanato-1-methoxypropane

  • MDL number MFCD18269656
  • Cas Number : 1340303-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-144800

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-144800
Availability details for
Quantity
  • Availability
  • Add to Cart