ENA432538353 Enamine

3-amino-4-sulfamoylbenzoic acid

  • MDL number MFCD24114563
  • Cas Number : 89694-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-147275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-147275
Availability details for
Quantity
  • Availability
  • Add to Cart