ENA432538360 Enamine

7-fluoro-6-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD18157741
  • Cas Number : 622369-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193026
Availability details for
Quantity
  • Availability
  • Add to Cart