ENA432538361 Enamine

7-fluoro-3-iodo-6-methoxyquinolin-4-ol

  • MDL number MFCD28139438
  • Cas Number : 1595523-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193027
Availability details for
Quantity
  • Availability
  • Add to Cart