ENA432538368 Enamine

(2-methylcyclopentyl)methanamine

  • MDL number MFCD11655618
  • Cas Number : 751464-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-151743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-151743
Availability details for
Quantity
  • Availability
  • Add to Cart