ENA432538370 Enamine

2-bromo-1-(1,2,3,4-tetrahydronaphthalen-1-yl)ethan-1-one

  • MDL number MFCD23802271
  • Cas Number : 1522497-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193052
Availability details for
Quantity
  • Availability
  • Add to Cart