ENA432538371 Enamine

2-propyldodecan-1-ol

  • MDL number MFCD21671982
  • Cas Number : 126147-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193054
Availability details for
Quantity
  • Availability
  • Add to Cart