ENA432538373 Enamine

4-chloro-7-fluoro-3-iodo-6-methoxyquinoline

  • MDL number MFCD28139442
  • Cas Number : 1602859-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193084
Availability details for
Quantity
  • Availability
  • Add to Cart