ENA432538374 Enamine

4-amino-8-bromo-7-methylquinoline-3-carbonitrile

  • MDL number MFCD24150850
  • Cas Number : 1513808-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193085
Availability details for
Quantity
  • Availability
  • Add to Cart