ENA432538376 Enamine

4-cyano-3-phenylbutanoic acid

  • MDL number MFCD20644552
  • Cas Number : 131906-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193087

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193087
Availability details for
Quantity
  • Availability
  • Add to Cart