ENA432538379 Enamine

6-(5-bromothiophen-2-yl)pyridazin-3-amine

  • MDL number MFCD16330198
  • Cas Number : 1226154-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193094
Availability details for
Quantity
  • Availability
  • Add to Cart