ENA432538385 Enamine

3-(dimethylamino)cyclohexane-1-carboxylic acid

  • MDL number MFCD20651194
  • Cas Number : 36268-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193112

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193112
Availability details for
Quantity
  • Availability
  • Add to Cart