ENA432538387 Enamine

2-(1,2,3,6-tetrahydropyridin-1-yl)ethan-1-amine

  • MDL number MFCD19206117
  • Cas Number : 27578-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193116
Availability details for
Quantity
  • Availability
  • Add to Cart