ENA432538388 Enamine

1-(6-bromo-3-fluoropyridin-2-yl)ethan-1-one

  • MDL number MFCD18258632
  • Cas Number : 1016228-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193117
Availability details for
Quantity
  • Availability
  • Add to Cart