ENA432538389 Enamine

1-(1-methanesulfonylcyclopropyl)methanamine hydrochloride

  • MDL number MFCD28055281
  • Cas Number : 1628734-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193118
Availability details for
Quantity
  • Availability
  • Add to Cart