ENA432538390 Enamine

2-(4-methyl-1,2,3,6-tetrahydropyridin-1-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD28139447
  • Cas Number : 34975-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193119
Availability details for
Quantity
  • Availability
  • Add to Cart