ENA432538400 Enamine

methyl 2-bromo-2-(thiophen-3-yl)acetate

  • MDL number MFCD28137600
  • Cas Number : 135660-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193150
Availability details for
Quantity
  • Availability
  • Add to Cart