ENA432538405 Enamine

1-methyl-3-phenyl-1H-1,2,4-triazol-5-amine

  • MDL number MFCD02853163
  • Cas Number : 3357-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193167
Availability details for
Quantity
  • Availability
  • Add to Cart