ENA432538406 Enamine

7-ethyl-[1,2,4]triazolo[4,3-c]pyrimidine-5-thiol

  • MDL number MFCD28139451
  • Cas Number : 1795069-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193168
Availability details for
Quantity
  • Availability
  • Add to Cart