ENA432538407 Enamine

2-propyl-1,3-oxazole-4-carbonitrile

  • MDL number MFCD28139452
  • Cas Number : 36190-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193169
Availability details for
Quantity
  • Availability
  • Add to Cart