ENA432538410 Enamine

2-phenyl-5H,6H,7H,8H-imidazo[1,2-a]pyrimidine hydrochloride

  • MDL number MFCD28139453
  • Cas Number : 1803571-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193177
Availability details for
Quantity
  • Availability
  • Add to Cart