ENA432538411 Enamine

4-(piperidin-4-yl)-4,5-dihydro-1H-1,2,4-triazol-5-one hydrochloride

  • MDL number MFCD28139454
  • Cas Number : 1795464-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193178
Availability details for
Quantity
  • Availability
  • Add to Cart