ENA432538416 Enamine

tert-butyl 4-hydroxy-4-(3-methoxyphenyl)piperidine-1-carboxylate

  • MDL number MFCD11849838
  • Cas Number : 871112-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156376
Availability details for
Quantity
  • Availability
  • Add to Cart